Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163018.1 | NP_001156490.1 | 10 - 11 | 1449 | 120 | |
NM_199196.2 | NP_954666.1 | 11 - 12 | 1518 | 120 | ||
GenBank mRNA | AK122213.1 | - | 11 - 12 | 1503 | 120 | |
AK135897.1 | - | 5 - 6 | 564 | 120 | ||
AK141264.1 | - | 2 - 3 | 122 | 120 | ||
BC039915.1 | - | 5 - 6 | 594 | 120 | ||
BC051099.1 | - | 2 - 3 | 212 | 120 | ||
BC064461.1 | - | 11 - 12 | 1518 | 6821922 | 120 | |
BC084591.1 | - | 6 - 7 | 789 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |