Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suz12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199196.2 | NP_954666.1 | 5 - 6 | 725 | 113 | |
GenBank mRNA | AK122213.1 | - | 5 - 6 | 710 | 113 | |
BC064461.1 | - | 5 - 6 | 725 | 6821922 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |