Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018814.3 | NP_061284.2 | 9 - 10 | 3071 | 149 | |
GenBank mRNA | AF096286.1 | - | 3 - 4 | 916 | 134 | |
AK129221.1 | - | 4 - 5 | 951 | 149 | ||
BC057333.1 | - | 2 - 3 | 829 | 134 | ||
BC066849.1 | - | 4 - 5 | 759 | 134 | ||
BC098370.1 | - | 4 - 5 | 676 | 134 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |