Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348447 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348448 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348446 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348449 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348450 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2019512 |
AK078782.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2019512 |
BC004631.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2019512 |
BC024371.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2019512 |
CX220128.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2019512 |
NM_013826.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
AK078782.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
BC024371.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
NM_013826.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
AF091872.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
AK002719.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
AK078782.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
BC024371.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
NM_013826.3 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
AF091872.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P674309 |
AK002719.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
168468 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
748383 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
714178 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
256059 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
475829 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505751 |
AK014391.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505751 |
AK032372.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505751 |
BC048151.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505751 |
BC058366.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505751 |
BC071201.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505751 |
NM_001037726.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505751 |
NM_009952.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
505751 |
NM_133828.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
665945 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P125046 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P135021 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
AK078782.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
BC024371.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
NM_013826.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
AF091872.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
AK002719.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
AK078782.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
BC024371.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
NM_013826.3 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
AF091872.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P674309 |
AK002719.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2019512 |
AK078782.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2019512 |
BC004631.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2019512 |
BC024371.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2019512 |
CX220128.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2019512 |
NM_013826.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P461675 |
AF091872.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348418 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348465 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348421 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348420 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348441 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348428 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348437 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348458 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348439 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348433 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348434 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348416 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348432 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348442 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348456 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348440 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348459 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348429 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348446 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348452 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348448 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348450 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348464 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348436 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348445 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348427 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348426 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348461 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348454 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348443 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348451 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348424 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348415 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348444 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348460 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348430 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P29348463 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P135021 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P125046 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P674309 |
AK078782.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
BC024371.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
NM_013826.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
AF091872.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
AK002719.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
AK078782.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
BC024371.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
NM_013826.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
AF091872.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P674309 |
AK002719.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P461675 |
AF091872.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P127212 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P125046 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P135021 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10870703 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10870707 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10870706 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10870704 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P10870702 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348448 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348446 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348449 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348447 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P29348450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348418 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348450 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348539 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348434 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348427 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348505 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348475 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348442 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348516 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348446 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348445 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348424 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348547 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348429 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348468 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348444 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348493 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348512 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348443 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348428 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348489 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348415 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348528 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348495 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348448 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348439 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348452 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348498 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348473 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348523 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348426 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348440 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348503 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348421 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348465 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348456 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348531 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348430 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348411 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348472 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P29348515 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348465 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348444 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348416 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348454 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348475 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348424 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348492 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348430 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348418 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10870704 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348448 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10846026 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348420 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10475407 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10505531 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348472 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348434 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348439 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348427 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10812193 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348546 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348547 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348476 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348450 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10620412 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348470 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348539 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348432 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348469 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348429 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10987628 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348484 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348538 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348486 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348456 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348466 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348426 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348428 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348464 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348534 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348482 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10799726 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348536 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348489 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348421 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348455 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348542 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348496 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348460 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10870697 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348436 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10476205 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348481 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10870698 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348478 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10870707 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348488 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348541 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348446 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348459 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348415 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348437 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348443 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P10987610 |
- |
- |
Agilent Six_ChIP_03_of_03_v2 |
A_68_P29348413 |
- |
- |