Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144524.2 | NP_653107.2 | 2 - 3 | 706 | 97 | |
GenBank mRNA | AK143549.1 | - | 2 - 3 | 707 | 97 | |
BC019148.1 | - | 2 - 3 | 721 | 5052974 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |