Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134042.2 | NP_598803.1 | 4 - 5 | 358 | 109 | |
GenBank mRNA | AF297860.1 | - | 4 - 5 | 358 | 109 | |
AK033587.1 | - | 4 - 5 | 385 | 109 | ||
AK082790.1 | - | 4 - 5 | 412 | 109 | ||
AK128924.1 | - | 4 - 5 | 390 | 109 | ||
AK147146.1 | - | 4 - 5 | 412 | 109 | ||
AK155814.1 | - | 4 - 5 | 409 | 109 | ||
AK164366.1 | - | 3 - 4 | 1958 | 109 | ||
AK169915.1 | - | 4 - 5 | 398 | 109 | ||
AK170305.1 | - | 4 - 5 | 411 | 109 | ||
AK171581.1 | - | 4 - 5 | 412 | 109 | ||
AK171896.1 | - | 4 - 5 | 401 | 109 | ||
BC031148.1 | - | 4 - 5 | 483 | 4975560 | 109 | |
BC033440.1 | - | 3 - 4 | 381 | 4212084 | 109 | |
BC115855.1 | - | 4 - 5 | 333 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |