Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134042.2 | NP_598803.1 | 7 - 8 | 862 | 67 | |
GenBank mRNA | AF297860.1 | - | 7 - 8 | 862 | 67 | |
AK033587.1 | - | 7 - 8 | 889 | 67 | ||
AK128924.1 | - | 7 - 8 | 894 | 67 | ||
AK147146.1 | - | 7 - 8 | 916 | 67 | ||
AK155814.1 | - | 7 - 8 | 913 | 67 | ||
AK169915.1 | - | 7 - 8 | 902 | 67 | ||
AK170305.1 | - | 7 - 8 | 915 | 67 | ||
AK171581.1 | - | 7 - 8 | 916 | 67 | ||
AK171896.1 | - | 7 - 8 | 905 | 67 | ||
BC031148.1 | - | 7 - 8 | 987 | 4975560 | 67 | |
BC033440.1 | - | 6 - 7 | 885 | 4212084 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |