Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028011.2 | NP_082287.1 | 9 - 10 | 958 | 85 | |
GenBank mRNA | AF395837.1 | - | 9 - 10 | 922 | 85 | |
AJ505007.1 | - | 9 - 10 | 919 | 85 | ||
AK009826.1 | - | 7 - 8 | 693 | 85 | ||
AK033712.1 | - | 9 - 10 | 960 | 85 | ||
AK083630.1 | - | 920 | 85 | |||
BC004710.1 | - | 2 - 3 | 1315 | 3499918 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |