Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145433.1 | NP_663408.2 | 2 - 3 | 779 | 99 | |
GenBank mRNA | AK156416.1 | - | 2 - 3 | 102 | ||
AK163986.1 | - | 2 - 3 | 771 | 99 | ||
AK164059.1 | - | 2 - 3 | 760 | 99 | ||
BC005625.1 | - | 2 - 3 | 779 | 3155732 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |