Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133865.2 | NP_598626.2 | 1 - 2 | 488 | 55 | |
GenBank mRNA | AK046571.1 | - | 1 - 2 | 491 | 55 | |
AK049115.1 | - | 1 - 2 | 483 | 55 | ||
AK051104.1 | - | 1 - 2 | 443 | 55 | ||
AK134324.1 | - | 1 - 2 | 248 | 55 | ||
AK137495.1 | - | 1 - 2 | 45 | 55 | ||
AK138568.1 | - | 1 - 2 | 546 | 55 | ||
AK154124.1 | - | 1 - 2 | 390 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons |