Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkiras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028024.2 | NP_082300.1 | 2 - 3 | 158 | 82 | |
GenBank mRNA | AK010371.1 | - | 2 - 3 | 158 | 82 | |
AK015918.1 | - | 163 | 82 | |||
AK052645.1 | - | 2 - 3 | 158 | 82 | ||
AK083475.1 | - | 2 - 3 | 201 | 82 | ||
AK149696.1 | - | 2 - 3 | 302 | 82 | ||
AK169878.1 | - | 2 - 3 | 183 | 82 | ||
BC013469.1 | - | 1 - 2 | 111 | 4219892 | 82 | |
BC024398.1 | - | 1 - 2 | 101 | 5003001 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |