Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rsbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172684.2 | NP_766272.2 | 3 - 4 | 1851 | 112 | |
GenBank mRNA | AB101658.1 | - | 3 - 4 | 1853 | 112 | |
AK087685.1 | - | 2 - 3 | 817 | 112 | ||
AK122577.1 | - | 3 - 4 | 1386 | 112 | ||
AK134093.1 | - | 4 - 5 | 970 | 112 | ||
AK138603.1 | - | 3 - 4 | 1395 | 112 | ||
BC094667.1 | - | 3 - 4 | 1376 | 112 | ||
BC139213.1 | - | 3 - 4 | 1820 | 8862235 | 112 | |
BC139215.1 | - | 3 - 4 | 1820 | 8862237 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |