Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289603.1 | NP_001276532.1 | 6 - 7 | 698 | 64 | |
NM_033476.3 | NP_258437.1 | 6 - 7 | 698 | 64 | ||
GenBank mRNA | AK131989.1 | - | 5 - 6 | 897 | 64 | |
AK146503.1 | - | 6 - 7 | 699 | 64 | ||
AK147898.1 | - | 5 - 6 | 907 | 64 | ||
AK158682.1 | - | 6 - 7 | 618 | 64 | ||
BC022131.1 | - | 6 - 7 | 671 | 5098336 | 64 | |
M84809.1 | - | 6 - 7 | 637 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |