Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700016572.mm.1 |
NM_001081160.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016572.mm.1 |
NM_001081160.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016572.mm.1 |
TC1700001794.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016569.mm.1 |
NM_001081160.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016569.mm.1 |
NM_001081160.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016569.mm.1 |
TC1700001794.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016571.mm.1 |
NM_001081160.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016571.mm.1 |
NM_001081160.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016571.mm.1 |
TC1700001794.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700016570.mm.1 |
NM_001081160.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016570.mm.1 |
NM_001081160.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700016570.mm.1 |
TC1700001794.mm.1 |
MTA TC |