Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033228.3 | NP_001028400.2 | 16 - 17 | 2585 | 120 | |
GenBank mRNA | AK035870.1 | - | 16 - 17 | 2586 | 120 | |
AK040830.1 | - | 7 - 8 | 1009 | 120 | ||
AK137169.1 | - | 16 - 17 | 2350 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |