Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026186.4 | NP_080462.2 | 7 - 8 | 1056 | 96 | |
GenBank mRNA | AK017849.1 | - | 7 - 8 | 996 | 96 | |
AK035158.1 | - | 7 - 8 | 985 | 96 | ||
BC021478.1 | - | 7 - 8 | 970 | 96 | ||
BC079553.1 | - | 7 - 8 | 978 | 6856335 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |