Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plxdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163608.1 | NP_001157080.1 | 5 - 6 | 744 | 74 | |
NM_028199.3 | NP_082475.3 | 5 - 6 | 723 | 74 | ||
GenBank mRNA | AF378760.1 | - | 5 - 6 | 657 | 74 | |
AK010361.1 | - | 5 - 6 | 723 | 74 | ||
AK036144.1 | - | 5 - 6 | 698 | 74 | ||
BC114352.2 | - | 5 - 6 | 643 | 40057410 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |