Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026072.1 | NP_080348.1 | 12 - 13 | 1365 | 124 | |
GenBank mRNA | AK014025.1 | - | 12 - 13 | 1357 | 124 | |
AK075902.1 | - | 12 - 13 | 1343 | 124 | ||
AK077779.1 | - | 12 - 13 | 1308 | 124 | ||
AK166770.1 | - | 12 - 13 | 1365 | 124 | ||
BC025437.1 | - | 12 - 13 | 1165 | 5025084 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |