Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081194.2 | NP_001074663.1 | 15 - 16 | 2828 | 72 | |
GenBank mRNA | BC150990.1 | - | 15 - 16 | 2836 | 9087903 | 72 |
BC150994.1 | - | 15 - 16 | 2832 | 9087907 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |