Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045527.1 | NP_001038992.1 | 4 - 5 | 1740 | 67 | |
GenBank mRNA | AK136195.1 | - | 1 - 2 | 206 | 67 | |
AK166561.1 | - | 1 - 2 | 182 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Mus musculus Probe spans exons |