Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK010261.1 | - | 1 - 2 | 44 | 64 | |
AK075843.1 | - | 1 - 2 | 43 | 64 | ||
AK131624.1 | - | 1 - 2 | 44 | 64 | ||
AK134921.1 | - | 1 - 2 | 44 | 64 | ||
AK148747.1 | - | 1 - 2 | 44 | 64 | ||
AK149111.1 | - | 1 - 2 | 46 | 64 | ||
AK150834.1 | - | 1 - 2 | 46 | 64 | ||
AK159105.1 | - | 1 - 2 | 44 | 64 | ||
AK159424.1 | - | 1 - 2 | 46 | 64 | ||
AK159517.1 | - | 1 - 2 | 44 | 64 | ||
BY772289.1 | - | 1 - 2 | 158 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mus musculus Inventoried Probe spans exons |