Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205343.1 | NP_001192272.1 | 24 - 25 | 3417 | 88 | |
NM_130887.3 | NP_570957.2 | 23 - 24 | 3351 | 88 | ||
GenBank mRNA | AK053198.1 | - | 24 - 25 | 3419 | 88 | |
AK086767.1 | - | 23 - 24 | 3334 | 88 | ||
BC005747.1 | - | 5 - 6 | 888 | 88 | ||
BC132475.1 | - | 23 - 24 | 3291 | 40130752 | 88 | |
BC137854.1 | - | 23 - 24 | 3291 | 8860873 | 88 | |
BC144975.1 | - | 24 - 25 | 3357 | 9053523 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons |