Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019579.3 | NP_062525.1 | 5 - 6 | 1061 | 78 | |
GenBank mRNA | AF199008.1 | - | 5 - 6 | 992 | 78 | |
AK014444.1 | - | 5 - 6 | 999 | 78 | ||
AK032731.1 | - | 5 - 6 | 1019 | 78 | ||
AK133187.1 | - | 5 - 6 | 937 | 78 | ||
AK162014.1 | - | 5 - 6 | 1019 | 78 | ||
BC067404.1 | - | 5 - 6 | 923 | 78 | ||
BC075698.1 | - | 5 - 6 | 1016 | 78 | ||
BC138624.1 | - | 5 - 6 | 940 | 8861644 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |