Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019579.3 | NP_062525.1 | 9 - 10 | 1636 | 88 | |
GenBank mRNA | AF199008.1 | - | 9 - 10 | 1567 | 88 | |
AK014444.1 | - | 9 - 10 | 88 | |||
AK032731.1 | - | 9 - 10 | 1594 | 88 | ||
AK133187.1 | - | 9 - 10 | 1529 | 88 | ||
AK162014.1 | - | 9 - 10 | 1594 | 88 | ||
BC067404.1 | - | 9 - 10 | 1497 | 88 | ||
BC075698.1 | - | 9 - 10 | 1591 | 88 | ||
BC138624.1 | - | 9 - 10 | 1515 | 8861644 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |