Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145478.2 | NP_663453.1 | 1 - 2 | 564 | 59 | |
GenBank mRNA | AK079623.1 | - | 1 - 2 | 284 | 59 | |
AK149478.1 | - | 1 - 2 | 284 | 59 | ||
AK159399.1 | - | 1 - 2 | 283 | 59 | ||
AK159600.1 | - | 1 - 2 | 283 | 59 | ||
AK159670.1 | - | 1 - 2 | 260 | 59 | ||
AK168014.1 | - | 1 - 2 | 573 | 59 | ||
AY026239.1 | - | 1 - 2 | 312 | 59 | ||
BC017621.1 | - | 1 - 2 | 564 | 4924687 | 59 | |
BC026639.1 | - | 1 - 2 | 253 | 4985551 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |