Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011767.2 | NP_035897.2 | 8 - 9 | 1577 | 116 | |
GenBank mRNA | AF071059.1 | - | 8 - 9 | 1575 | 116 | |
AK034963.1 | - | 1 - 2 | 146 | 116 | ||
BC094220.1 | - | 7 - 8 | 1465 | 115 | ||
BC141139.1 | - | 8 - 9 | 1561 | 9055751 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |