Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spop | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025287.2 | NP_079563.2 | 5 - 6 | 541 | 63 | |
GenBank mRNA | AB071989.1 | - | 3 - 4 | 350 | 63 | |
AK004669.1 | - | 5 - 6 | 511 | 63 | ||
AK028201.1 | - | 4 - 5 | 677 | 63 | ||
AK030746.1 | - | 4 - 5 | 728 | 63 | ||
AK132383.1 | - | 4 - 5 | 717 | 63 | ||
AK159482.1 | - | 4 - 5 | 694 | 63 | ||
AK160858.1 | - | 4 - 5 | 639 | 63 | ||
AK166581.1 | - | 4 - 5 | 651 | 63 | ||
AK168185.1 | - | 4 - 5 | 663 | 63 | ||
AK168546.1 | - | 4 - 5 | 730 | 63 | ||
AY538613.1 | - | 3 - 4 | 353 | 63 | ||
BC043131.1 | - | 4 - 5 | 560 | 6811159 | 63 | |
BC045205.1 | - | 5 - 6 | 511 | 4988521 | 63 | |
BY726358.1 | - | 5 - 6 | 542 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |