Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfrsf1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011610.3 | NP_035740.2 | 1 - 2 | 146 | 62 | |
GenBank mRNA | AK004752.1 | - | 1 - 2 | 147 | 62 | |
AK004844.2 | - | 1 - 2 | 146 | 62 | ||
AK140332.1 | - | 1 - 2 | 147 | 62 | ||
AK153301.1 | - | 1 - 2 | 147 | 62 | ||
AK155385.1 | - | 1 - 2 | 147 | 62 | ||
BC141405.1 | - | 1 - 2 | 133 | 9056018 | 62 | |
M59378.1 | - | 1 - 2 | 119 | 62 | ||
M60469.1 | - | 1 - 2 | 117 | 62 | ||
X76401.1 | - | 1 - 2 | 32 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |