Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016706.2 | NP_057915.2 | 2 - 3 | 1365 | 114 | |
GenBank mRNA | AF208663.1 | - | 2 - 3 | 1347 | 114 | |
BC031167.1 | - | 2 - 3 | 1380 | 4983919 | 114 | |
CF105200.1 | - | 1 - 2 | 163 | 114 | ||
CF949734.1 | - | 1 - 2 | 639 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |