Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poldip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026389.3 | NP_080665.1 | 1 - 2 | 176 | 61 | |
GenBank mRNA | AK196491.1 | - | 1 - 2 | 39 | 61 | |
BC006833.1 | - | 1 - 2 | 176 | 3597851 | 61 | |
BC012879.1 | - | 1 - 2 | 188 | 4166791 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |