Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008702.3 | NP_032728.3 | 6 - 7 | 2211 | 59 | |
GenBank mRNA | AF036332.1 | - | 6 - 7 | 1338 | 59 | |
AK078118.1 | - | 6 - 7 | 1320 | 59 | ||
BC057667.1 | - | 6 - 7 | 1296 | 4235238 | 59 | |
BC058652.1 | - | 6 - 7 | 2159 | 6406851 | 59 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |