Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sele | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011345.2 | NP_035475.1 | 12 - 13 | 1915 | 108 | |
GenBank mRNA | AK153623.1 | - | 12 - 13 | 1934 | 108 | |
BC119083.1 | - | 12 - 13 | 1866 | 8733832 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |