Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpine3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199945.1 | NP_001186874.1 | 7 - 8 | 1139 | 95 | |
GenBank mRNA | AK053602.1 | - | 7 - 8 | 1141 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |