Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010778.3 | NP_034908.1 | 6 - 7 | 913 | 76 | |
GenBank mRNA | AB001566.1 | - | 6 - 7 | 890 | 76 | |
AB010919.1 | - | 6 - 7 | 910 | 76 | ||
AK006632.1 | - | 6 - 7 | 914 | 76 | ||
BC150660.1 | - | 6 - 7 | 914 | 9087571 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |