Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ero1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015774.3 | NP_056589.1 | 12 - 13 | 1123 | 130 | |
GenBank mRNA | AK009667.1 | - | 2 - 3 | 348 | 130 | |
AK142595.1 | - | 12 - 13 | 1114 | 130 | ||
BC025102.1 | - | 12 - 13 | 1075 | 2654098 | 130 | |
CT010195.1 | - | 12 - 13 | 1041 | 130 | ||
CT010259.1 | - | 12 - 13 | 1041 | 130 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |