Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037221.2 | NP_001032298.1 | 10 - 11 | 2522 | 79 | |
NM_001163433.1 | NP_001156905.1 | 9 - 10 | 2002 | 79 | ||
NM_028966.3 | NP_083242.1 | 9 - 10 | 2258 | 79 | ||
GenBank mRNA | AK017081.1 | - | 5 - 6 | 836 | 79 | |
AK034323.1 | - | 8 - 9 | 1852 | 79 | ||
AK133470.1 | - | 9 - 10 | 2237 | 80 | ||
AK134243.1 | - | 10 - 11 | 2520 | 79 | ||
AK161277.1 | - | 9 - 10 | 1996 | 79 | ||
BC049740.1 | - | 5 - 6 | 808 | 79 | ||
BC145827.1 | - | 9 - 10 | 2124 | 40131130 | 79 | |
BC145829.1 | - | 9 - 10 | 2123 | 40131132 | 79 | |
DQ278487.1 | - | 9 - 10 | 2035 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |