Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081363.2 | NP_001074832.2 | 5 - 6 | 707 | 75 | |
GenBank mRNA | AK080758.1 | - | 5 - 6 | 694 | 75 | |
AK165236.1 | - | 5 - 6 | 708 | 75 | ||
DQ642022.1 | - | 4 - 5 | 564 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |