Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mark1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145515.2 | NP_663490.2 | 11 - 12 | 1194 | 133 | |
GenBank mRNA | AB185282.1 | - | 11 - 12 | 1117 | 133 | |
AK173181.1 | - | 10 - 11 | 1044 | 133 | ||
BC112400.1 | - | 11 - 12 | 1123 | 40059725 | 133 | |
CX210969.1 | - | 3 - 4 | 254 | 133 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |