Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnase1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007870.3 | NP_031896.3 | 1 - 2 | 356 | 96 | |
GenBank mRNA | AF047355.1 | - | 1 - 2 | 331 | 96 | |
AK156109.1 | - | 189 | 96 | |||
AK171843.1 | - | 1 - 2 | 189 | 96 | ||
AK172254.1 | - | 1 - 2 | 749 | 96 | ||
BC012671.1 | - | 1 - 2 | 309 | 4160709 | 96 | |
U76110.1 | - | 1 - 2 | 165 | 96 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |