Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dbt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010022.3 | NP_034152.2 | 9 - 10 | 1272 | 79 | |
GenBank mRNA | AK165959.1 | - | 9 - 10 | 1282 | 79 | |
BC055890.1 | - | 9 - 10 | 1228 | 4242586 | 79 | |
L42996.1 | - | 9 - 10 | 1225 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |