Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubxn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145441.3 | NP_663416.1 | 4 - 5 | 472 | 143 | |
GenBank mRNA | AK031821.1 | - | 4 - 5 | 473 | 143 | |
AK041556.1 | - | 4 - 5 | 509 | 143 | ||
AK134666.1 | - | 4 - 5 | 505 | 143 | ||
BC004632.1 | - | 4 - 5 | 424 | 3585699 | 143 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |