Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093752.1 | NP_001087221.1 | 1 - 2 | 127 | 59 | |
GenBank mRNA | AK011472.1 | - | 1 - 2 | 65 | 59 | |
AK040467.1 | - | 1 - 2 | 102 | 59 | ||
AK163152.1 | - | 1 - 2 | 95 | 59 | ||
BC037053.1 | - | 1 - 2 | 50 | 59 | ||
BC038928.1 | - | 1 - 2 | 86 | 59 | ||
BC055763.1 | - | 1 - 2 | 73 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |