Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdlim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190852.1 | NP_001177781.1 | 13 - 14 | 1666 | 82 | |
NM_001190854.1 | NP_001177783.1 | 10 - 11 | 1270 | 82 | ||
NM_001190855.1 | NP_001177784.1 | 11 - 12 | 1402 | 82 | ||
NM_001190856.1 | NP_001177785.1 | 12 - 13 | 1546 | 82 | ||
NM_019808.3 | NP_062782.2 | 10 - 11 | 1597 | 82 | ||
GenBank mRNA | AK089779.1 | - | 10 - 11 | 1607 | 82 | |
AK147999.1 | - | 10 - 11 | 1537 | 82 | ||
BC020145.1 | - | 2 - 3 | 226 | 82 | ||
BC037476.1 | - | 10 - 11 | 1561 | 4457868 | 82 | |
HM369794.1 | - | 9 - 10 | 1117 | 82 | ||
HM369795.1 | - | 10 - 11 | 1249 | 82 | ||
HM369796.1 | - | 11 - 12 | 1393 | 82 | ||
HM369797.1 | - | 12 - 13 | 1513 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |