Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rundc3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198620.1 | NP_941022.1 | 5 - 6 | 893 | 71 | |
GenBank mRNA | BC057375.1 | - | 1 - 2 | 106 | 71 | |
BC058808.1 | - | 5 - 6 | 893 | 6491326 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |