Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clasrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016680.5 | NP_057889.3 | 17 - 18 | 1898 | 70 | |
NR_125832.1 | 17 - 18 | 1902 | 70 | |||
NR_125833.1 | 16 - 17 | 2135 | 70 | |||
NR_125834.1 | 16 - 17 | 2151 | 70 | |||
NR_125835.1 | 16 - 17 | 2113 | 70 | |||
NR_125836.1 | 14 - 15 | 2015 | 70 | |||
GenBank mRNA | AB080582.1 | - | 16 - 17 | 1761 | 70 | |
AB080583.1 | - | 16 - 17 | 1765 | 70 | ||
AF042799.1 | - | 16 - 17 | 2116 | 70 | ||
AF325456.1 | - | 7 - 8 | 1744 | 70 | ||
AK003261.2 | - | 5 - 6 | 612 | 70 | ||
AK009334.1 | - | 16 - 17 | 2102 | 70 | ||
AK159443.1 | - | 17 - 18 | 1871 | 70 | ||
BC010728.1 | - | 17 - 18 | 1850 | 70 | ||
BC037756.1 | - | 2 - 3 | 235 | 70 | ||
BC051916.1 | - | 17 - 18 | 1877 | 5707517 | 70 | |
BC060092.1 | - | 16 - 17 | 4266 | 70 | ||
BC103781.1 | - | 14 - 15 | 1981 | 70 | ||
BC106129.1 | - | 16 - 17 | 2114 | 70 | ||
BY591198.1 | - | 2 - 3 | 171 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |