Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013648.6 | NP_038676.1 | 2 - 3 | 282 | 70 | |
GenBank mRNA | AF038538.1 | - | 2 - 3 | 238 | 70 | |
AK144271.1 | - | 2 - 3 | 245 | 70 | ||
AK165915.1 | - | 2 - 3 | 272 | 70 | ||
AU066588.1 | - | 2 - 3 | 283 | 70 | ||
BC031370.1 | - | 2 - 3 | 239 | 5359358 | 70 | |
CJ176292.1 | - | 2 - 3 | 249 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |