Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gzmc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010371.2 | NP_034501.2 | 2 - 3 | 278 | 91 | |
GenBank mRNA | BC120755.1 | - | 2 - 3 | 228 | 40129678 | 91 |
BC120757.1 | - | 2 - 3 | 228 | 40129680 | 91 | |
BC145030.1 | - | 2 - 3 | 228 | 9053578 | 91 | |
X12822.1 | - | 2 - 3 | 222 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protease Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |