Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400015522.mm.1 |
NM_199470.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015522.mm.1 |
NM_199470.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015522.mm.1 |
TC1400002062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015524.mm.1 |
NM_199470.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015524.mm.1 |
NM_199470.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015524.mm.1 |
TC1400002062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015525.mm.1 |
NM_199470.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015525.mm.1 |
NM_199470.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015525.mm.1 |
TC1400002062.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400015523.mm.1 |
NM_199470.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015523.mm.1 |
NM_199470.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400015523.mm.1 |
TC1400002062.mm.1 |
MTA TC |