Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sec11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019951.1 | NP_064335.1 | 4 - 5 | 525 | 78 | |
GenBank mRNA | AB025405.1 | - | 4 - 5 | 525 | 78 | |
AK007449.1 | - | 4 - 5 | 523 | 78 | ||
BC010484.1 | - | 4 - 5 | 517 | 3483051 | 78 | |
BC057885.1 | - | 4 - 5 | 717 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |