Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028026.2 | NP_082302.2 | 4 - 5 | 331 | 115 | |
NR_027976.1 | 3 - 4 | 804 | 115 | |||
GenBank mRNA | AK004708.1 | - | 3 - 4 | 769 | 115 | |
AK010448.1 | - | 4 - 5 | 297 | 115 | ||
AK034740.1 | - | 4 - 5 | 295 | 115 | ||
AK049132.1 | - | 4 - 5 | 329 | 115 | ||
AK169377.1 | - | 3 - 4 | 768 | 115 | ||
AK169527.1 | - | 4 - 5 | 331 | 115 | ||
AK220207.1 | - | 4 - 5 | 300 | 115 | ||
BC028870.1 | - | 4 - 5 | 284 | 4037614 | 115 | |
BC068122.1 | - | 3 - 4 | 758 | 115 | ||
BC087940.1 | - | 4 - 5 | 290 | 30302907 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |